Learn More
Galectin 8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534861
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: U87-MG, SK-N-SH, SK-N-AS, HepG2, human LGALS8-transfected 293T, LGALS8 transfected 293T lysate. Predicted reactivity: Human (99%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Galectin 8 Polyclonal antibody specifically detects Galectin 8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
Galectin 8 | |
Polyclonal | |
Unconjugated | |
Lgals8 | |
1200015E08Rik; 30 kDa S-type lectin; AI326142; D13Ertd524e; Gal-8; galectin 8; galectin-8; galectin-8g; galectin-8L; lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 8; lectin, galactoside binding soluble 8; lectin, galactoside-binding, soluble, 8; LGALS8; Lgals-8; PCTA1; PCTA-1; Po66 carbohydrate binding protein; po66 carbohydrate-binding protein; Po66-CBP; prostate carcinoma tumor antigen 1; RL-30; RP11-385F5.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3964 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.68 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O00214 | |
Lgals8 | |
Recombinant protein fragment contains a sequence corresponding to a region within amino acids 39 and 268 of Galectin 8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |