missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAGE12I Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA546157
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: AGQGPKPEAH SQEQGHPQTG CECEDGPDGQ EMDPPNPEEV KTPEEGEKQS Sequence homology: Human: 100%
GAGE12I Polyclonal antibody specifically detects GAGE12I in Human samples. It is validated for Western Blot.Especificaciones
GAGE12I | |
Polyclonal | |
Unconjugated | |
GAGE12I | |
AL4; cancer/testis antigen 4.7; CT4.7; G antigen 12F; g antigen 12F/G/I; G antigen 12G; G antigen 12I; G antigen 7; G antigen 7B; GAGE12F; GAGE-12F; GAGE12G; GAGE-12G; GAGE12I; GAGE-12I; GAGE7; GAGE-7; GAGE7B; GAGE-7B; GAGE-8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26748 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P0CL82 | |
GAGE12I | |
Synthetic peptide directed towards the C-terminal of human GAGE12I | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |