missing translation for 'onlineSavingsMsg'
Learn More

GABPB2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA531414

Detalles adicionales : Peso : 0.01000kg

Código de producto. 13465937

  • 473.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (83%), Chimpanzee (98%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GABPB2 Polyclonal antibody specifically detects GABPB2 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

GABPB2
Polyclonal
Unconjugated
Gabpb2
1810015F01Rik; 5830427M07; 9430006E19Rik; A430024B14Rik; AV050852; BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4TF1B; GA binding protein transcription factor beta subunit 1; GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor beta subunit 2; GA binding protein transcription factor subunit beta 2; GA binding protein transcription factor, alpha subunit pseudogene; GA binding protein transcription factor, beta subunit 1; GA binding protein transcription factor, beta subunit 2; GA repeat binding protein, beta 2; GA-binding protein beta-2-1; GA-binding protein subunit beta-1; GA-binding protein subunit beta-2; GABP subunit beta-1; GABP subunit beta-2; GABP subunit beta-2-1; GABP2; GABPAP; GABPB; GABPB1; GABPB-1; Gabpb2; GABPB-2; Gabpb2-1; LOW QUALITY PROTEIN: GA-binding protein subunit beta-2; NRF2B1; NRF2B2; Nuclear respiratory factor 2; RGD1565234; RP11-68I18.1; Transcription factor E4TF1-47; transcription factor E4TF1-53
Rabbit
Antigen affinity chromatography
RUO
126626
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.48 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q8TAK5
Gabpb2
Recombinant protein fragment corresponding to a region within amino acids 209 and 395 of Human GABPB2
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales