Learn More
GABARAPL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA527594
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: Jurkat, Raji, NCI-H929. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (97%), Xenopus laevis (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GABARAPL2 Polyclonal antibody specifically detects GABARAPL2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
GABARAPL2 | |
Polyclonal | |
Unconjugated | |
GABARAPL2 | |
0610012F20Rik; 2900019O08Rik; AI173605; ATG8; ATG8C; FLC3A; GABA type A receptor associated protein like 2; GABA(A) receptor-associated protein like 2; GABA(A) receptor-associated protein-like 2; gabarapl2; gamma-aminobutyric acid (GABA) A receptor-associated protein-like 2; gamma-aminobutyric acid (GABA-A) receptor-associated protein-like 2; gamma-aminobutyric acid receptor-associated protein-like 2; Ganglioside expression factor 2; GATE16; GATE-16; Gef2; GEF-2; Genera; general protein transport factor p16; Golgi-associated ATPase enhancer of 16 kDa; MAP1 light chain 3 related protein; MAP1 light chain 3-related protein; zgc:92319 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11345, 64670, 93739 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
P60520, P60521, P60522 | |
GABARAPL2 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 104 of Human GABARAPL2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |