Learn More
GABRA6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen OPA104114
Detalles adicionales : Peso : 0.01000kg
Descripción
In Western blot, this antibody detects a ∽57 kDa protein representing GABA-A R alpha 6 subunit in rat brain lysate. The immunogenic sequence has no homology with any other GABA-A receptor subunits.
GABRA6 Polyclonal antibody specifically detects GABRA6 in Mouse, Rat samples. It is validated for Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Especificaciones
GABRA6 | |
Polyclonal | |
Unconjugated | |
Gabra6 | |
alpha6; GABA subunit A receptor alpha 6; GABA(A) receptor subunit alpha-6; GABA(A) receptor, alpha 6; GABA-A receptor alpha-6 subunit; GABAAR alpha6; GABA-ARalpha6; GABRA 6; Gabra6; Gabra-6; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 6; gamma-aminobutyric acid (GABA) A receptor, alpha 6; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 6; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 6; gamma-aminobutyric acid A receptor, alpha 6; gamma-aminobutyric acid receptor subunit alpha-6; gamma-aminobutyric acid type A receptor alpha6 subunit; subunit alpha 6 gamma-aminobutyric acid (GABA-A) receptor | |
Rabbit | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
P16305, P30191 | |
Gabra6 | |
Synthetic peptide corresponding to amino acid residues specific to the alpha 6 subunit of rat GABRA6 conjugated to KLH | |
RUO | |
14399, 29708 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |