Learn More
FABP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca:  Invitrogen PA513461
Detalles adicionales : Peso : 0.01000kg
Código de producto. 10372280
  • 472.00€ / 100 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock
Descripción
Descripción
This antibody is predicted to react with bovine, porcine and rat based on sequence homology.
FABP3 Polyclonal antibody specifically detects FABP3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
Especificaciones
Especificaciones
FABP3
Polyclonal
Unconjugated
FABP3
422 protein; Cardiac Fatty Acid Binding Protein; FABP 3; FABP11; Fabp3; Fabph1; Fabph-1; Fabph4; Fabph-4; fatty acid binding protein 11; Fatty acid binding protein 11 (FABP11); fatty acid binding protein 3; Fatty acid binding protein 3 heart; fatty acid binding protein 3, muscle and heart; Fatty acid binding protein heart; fatty acid binding protein heart 1; fatty acid binding protein heart 4; fatty acid-binding protein 3; Fatty acid-binding protein, heart; heart FABP; heart fatty acid binding protein; Heart type fatty acid binding protein; heart-type fatty acid-binding protein; H-FABP; M FABP; Mammary derived growth inhibitor; Mammary-derived growth inhibitor; MDGI; M-FABP; muscle and heart (mammary derived growth inhibitor); Muscle fatty acid binding protein; Muscle fatty acid-binding protein; Muscle O-FABP; Mylein protein P2 homolog / adopocyte; O-FABP
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
14077, 2170
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P05413, P11404
FABP3
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human FABP3
400 μL
Primary
Human, Mouse