missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDB1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA19036
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with bovine, canine and rat based on sequence homology.
DDB1 Polyclonal antibody specifically detects DDB1 in Human, Mouse samples. It is validated for Western Blot.Especificaciones
DDB1 | |
Polyclonal | |
Unconjugated | |
DDB1 | |
127kDa; AA408517; damage specific DNA binding protein 1; damaged-DNA recognition protein 1; damage-specific DNA binding protein 1, 127kDa; damage-specific DNA-binding protein 1; damage-specific DNA-binding protein, DNA repair; DDB p127 subunit; DDB1; DDBa; DNA damage-binding protein 1; DNA damage-binding protein a; DNA repair protein; HBV X-associated protein 1; p127-Ddb1; UV-damaged DNA-binding factor; UV-damaged DNA-binding protein 1; UV-DDB 1; UV-DDB1; XAP1; XAP-1; xeroderma pigmento; Xeroderma pigmentosum group E-complementing protein; XPCE; XPE; XPE-BF; XPE-binding factor | |
Goat | |
Antigen affinity chromatography | |
RUO | |
13194, 1642 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q16531, Q3U1J4 | |
DDB1 | |
Synthetic peptide corresponding to the C terminus amino acids DLIKVVEELTRIH. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |