Learn More
Cyclin E Mouse anti-Human, Unlabeled, Clone: HE12, BD
Mouse Monoclonal Antibody
329.00€ - 576.00€
Especificaciones
Antígeno | Cyclin E |
---|---|
Clon | HE12 |
Aplicaciones | Western Blot |
Clasificación | Monoclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
10537085
|
BD Biosciences
551159 |
50 μg |
329.00€
cada uno |
Fecha estimada de envío: 16-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15876558
|
BD Biosciences
551160 |
150 μg |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
Descripción
Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins (regulatory subunits) bind to cdks (catalytic subunits) to form complexes that regulate the progression of the cell cycle. These complexes are regulated by activating and inhibitory phosphorylation events, as well as by interactions with small proteins that bind to cyclins, cdks, or cyclin-cdk complexes, e.g., p21 and p27 [Kip1]. Cyclin E is expressed in G1 and associates with cdk2 to form an active kinase where it plays an important role in the regulation of the G1/S restriction checkpoint in the cell cycle. Abberant expression of cyclin E has been reported to be associated with the oncogenic transformation of cells. This antibody has been reported not to cross-react with mouse cyclin E.
Especificaciones
Cyclin E | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Primary | |
Store undiluted at -20°C. |
HE12 | |
Monoclonal | |
Mouse | |
Cell Biology | |
IgG1 | |
Affinity Purified |
For Research Use Only.