missing translation for 'onlineSavingsMsg'
Learn More

Csk Mouse, Unlabeled, Clone: 52, BD

Mouse Monoclonal Antibody

459.19€

Especificaciones

Antígeno Csk
Clon 52
Concentración 250μg/mL
Aplicaciones Western Blot
Clasificación Monoclonal
Ver más especificaciones

Productos 1
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15835249
Ver documentos Detalles de la promoción
BD Biosciences
610080
150 μg Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A Ver productos alternativos
Ahorre hasta 
»
Descripción

Descripción

The csk gene encodes a protein kinase that phosphorylates several Src family kinases including p60(c-src), p59(fyn), p53/56(lyn), p56(lck), and p62(yes). This phosphorylation occurs specifically at the carboxy-terminal tyrosine, Y527. Csk (c-src kinase) was identified based on its ability to phosphorylate exogenous Src in mammalian src-deficient cells. Establishment of a mouse csk- cell line revealed that phosphorylation of Y527 in Src was reduced 20-50%. This correlated with an increase of Src kinase activity, thus indicating Csk is a negative regulator of src. Csk-deficient cells also show an elevation in the kinase activity of p59(fyn) and p53/56(lyn). These data suggest that Csk may be a universal negative regulator of Src family kinases in vivo.

Host Species: Mouse
Clone: 52
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Rat Csk aa. 1-156
Formula Weight [Chemical]: 50kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Especificaciones

Especificaciones

Csk
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
52
Western Blot
Unconjugated
RUO
Canine, Chicken, Human, Mouse, Rat
Rat Csk aa. 1-156
Primary
Store undiluted at -20°C.
Videos
SDS
Documentos

Documentos

Product Certifications

For Research Use Only.