missing translation for 'onlineSavingsMsg'
Learn More

CLK1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA538617

Detalles adicionales : Peso : 0.00900kg

Código de producto. 15575531

  • 470.00€ / 100 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases. In the nucleus, the encoded protein phosphorylates serine/arginine-rich proteins involved in pre-mRNA processing, releasing them into the nucleoplasm. The choice of splice sites during pre-mRNA processing may be regulated by the concentration of transacting factors, including serine/arginine rich proteins. Therefore, the encoded protein may play an indirect role in governing splice site selection.
Especificaciones

Especificaciones

CLK1
Polyclonal
Unconjugated
CLK1
CLK, CLK/STY, STY, CDC28/CDC2-like kinase, dual specificity protein kinase CLK1, protein tyrosine kinase STY
Rabbit
Antigen affinity chromatography
RUO
1195, 12747
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
P49759, P22518
CLK1
A synthetic peptide derived from the internal region of human CLK1
100 μg
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales