missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p19ARF Monoclonal Antibody (5-C3-1), Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen MA116665
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody does not react with human or Golden hamster. Armenian hamster has not been tested.
p19ARF Monoclonal antibody specifically detects p19ARF in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Especificaciones
p19ARF | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P42771, Q64364, Q8QZZ9 | |
Cdkn2a | |
Synthetic peptide containing amino acids 54-75 of the murine p19ARF. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
5-C3-1 | |
Unconjugated | |
Cdkn2a | |
Alternative reading frame; Arf; ARF-INK4a; CDK4 inhibitor p16-INK4; CDK4I; CDKN2; CDKN2A; cdkn2a {ECO:0000312; cell cycle inhibitor; cell cycle negative regulator beta; cell cycle regulator; CMM2; cyclin dependent kinase inhibitor 2A; Cyclin dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2a p16Ink4a; cyclin-dependent kinase inhibitor 2a p19Arf; cyclin-dependent kinase inhibitor 2A, isoform 1; cyclin-dependent kinase inhibitor 2A, isoform 2; cyclin-dependent kinase inhibitor 2A, isoform 3; cyclin-dependent kinase inhibitor 2A, isoforms 1/2; cyclin-dependent kinase inhibitor 2A, isoforms 1/2/3-like; cyclin-dependent kinase inhibitor protein; EMBL:AAL76336.1, ECO:0000312; EMBL:AAL76338.1, ECO:0000312; INK4; INK4A; Ink4a/Arf; INK4a-ARF; LOC100764983; | |
Rat | |
Protein A | |
RUO | |
1029, 12578, 25163 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |