Learn More
CD86 Monoclonal Antibody (BU63), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA181186
Detalles adicionales : Peso : 0.01000kg
Descripción
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD86 antibody, clone Bu63 recognizes human CD86 also known as B7-2, a type I transmembrane protein expressed by monocytes and activated B cells.
CD86 Monoclonal antibody specifically detects CD86 in Human samples. It is validated for Flow Cytometry.Especificaciones
CD86 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide; pH 7.2 | |
P42081 | |
CD86 | |
Human peripheral blood lymphocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
BU63 | |
FITC | |
CD86 | |
activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2 | |
Mouse | |
Protein A | |
RUO | |
942 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |