missing translation for 'onlineSavingsMsg'
Learn More

CD84 Monoclonal Antibody (CD84.1.21), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA134024

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11594053

  • 614.00€ / 200 microgramos
En stock
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CD84 Monoclonal antibody specifically detects CD84 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.
Especificaciones

Especificaciones

CD84
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
Q9UIB8
CD84
Mouse pre-B cell line 300.19 transfected with CD84 cDNA.
200 μg
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunoprecipitation
CD84.1.21
Unconjugated
CD84
A130013D22Rik; CD84; CD84 antigen; CD84 antigen (leukocyte antigen); CD84 molecule; CDw84; cell surface antigen MAX.3; DKFZp781E2378; hCD84; Hly9-beta; leucocyte differentiation antigen CD84; leukocyte antigen CD84; leukocyte differentiation antigen CD84; LY9B; mCD84; RGD1564553; Signaling lymphocytic activation molecule 5; SLAM family member 5; SLAMF5
Mouse
Protein G
RUO
8832
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales