missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80 Mouse anti-Rat, APC, Clone: 3H5, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MR6505
Detalles adicionales : Peso : 0.50000kg
Descripción
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28 and of an inhibitory receptor CTLA-4 (CD152). The both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, the both molecules can also substitute for each other in this process. The question what are the differences in CD80 and CD86 competency has not been fully elucidated yet; there are still conflicts in results about their respective roles in initiation or sustaining of the T cell immune response.Especificaciones
CD80 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide | |
B7-1, B7.1, Cd28l, Ly-53, Ly53, MIC17, TS/A-1, LAB7, B7.1, CD28LG, CD28LG1 | |
Mouse | |
Purified | |
RUO | |
25408 | |
4° C | |
Liquid |
Flow Cytometry | |
3H5 | |
APC | |
Cd80 | |
Cd80 | |
Rat CD80. | |
500 μL | |
Primary | |
Rat | |
Antibody | |
IgG1 κ |