missing translation for 'onlineSavingsMsg'
Learn More

CD79a Monoclonal Antibody (SP18), Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Invitrogen MA516344

Detalles adicionales : Peso : 0.01000kg

Código de producto. 13431077

  • 514.00€ / 500 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil or B cell lymphoma.

CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.
Especificaciones

Especificaciones

CD79a
Monoclonal
Unconjugated
P11912
CD79A
Synthetic peptide derived from N-terminus region of human CD79a protein.
RUO
973
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin)
SP18
CD79A
IGA, MB-1, B-cell antigen receptor complex-associated protein alpha chain, CD79a antigen immunoglobulin-associated alpha, MB-1 membrane glycoprotein
Rabbit
500 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales