Learn More
CD79a Monoclonal Antibody (HM57), PE, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA516711
Detalles adicionales : Peso : 0.01000kg
Descripción
Membrane permeabilization is required for flow cytometry applications. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. Once reconstituted, this product has a shelf life of approx. 18 months. Mouse anti Human CD79a antibody, clone HM57 recognizes an epitope within the cytoplasmic domain of CD79a.
CD79a Monoclonal antibody specifically detects CD79a in Human, Many, Mouse, Rat samples. It is validated for Flow Cytometry.Especificaciones
CD79a | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P11911, P11912 | |
CD79A | |
Synthetic peptide corresponding to 202-216 amino acid sequence of human mb-1. | |
100 Tests | |
Primary | |
Human, Multi-species, Mouse, Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
HM57 | |
PE | |
CD79A | |
B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein | |
Mouse | |
Protein A | |
RUO | |
100913063, 12518, 973 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |