missing translation for 'onlineSavingsMsg'
Learn More

P-Selectin Monoclonal Antibody (AK-6), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA516567

353.35 EUR hasta el 2024-03-29
Use el código promocional "21615" para aplicar la oferta.



Detalles adicionales : Peso : 0.01000kg

Código de producto. 15265849

  • 530.00€ / 200 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. A suggested positive control for immunohistochemical applications is tonsil. This product does not require antigen retrieval using heat treatment prior to staining of paraffin sections but staining may be enhanced using a sodium citrate buffer, pH6.0. Mouse anti Human CD62P antibody, clone AK-6 recognizes the CD62P, also known as P-selectin, Granule membrane protein 140, GMP140, Leukocyte-endothelial cell adhesion molecule 3 or Platelet activation dependent granule-external membrane protein.

P-Selectin Monoclonal antibody specifically detects P-Selectin in Human, Rhesus Monkey samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

P-Selectin
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P16109
SELP
Human platelet membrane glycoproteins
200 μg
Primary
Human, Rhesus Monkey
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
AK-6
Unconjugated
SELP
CD62; CD62 antigen-like family member P; CD62P; CD62P antigen; CD62P; CD62 homolog; FLJ45155; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; Grmp; LECAM3; leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; PSELECT; P-selectin; P-selectin precursor; RP1-86F14.2; selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); selectin, platelet; Selp
Mouse
Protein A
RUO
6403, 701277
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales