Learn More
CD61 Monoclonal Antibody (HM beta 3.1), PE, Invitrogen™
Armenian Hamster Monoclonal Antibody
Marca: Invitrogen MA516709
Detalles adicionales : Peso : 0.01000kg
Descripción
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD61 Monoclonal antibody specifically detects CD61 in Mouse, Rat samples. It is validated for Flow Cytometry.Especificaciones
CD61 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
O54890 | |
ITGB3 | |
Mouse alpha 5 beta 3 protein purified from the mouse hybridoma 2B4 | |
100 Tests | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry | |
HM beta 3.1 | |
PE | |
ITGB3 | |
antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa | |
Armenian Hamster | |
Protein G | |
RUO | |
16416, 29302 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |