Learn More
CD40 Mouse anti-Human, NA/LE, Unlabeled, Clone: 5C3, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 555586
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody reacts with a 45-48kDa type I integral membrane glycoprotein present on peripheral blood and tonsillar B cells, but not expressed on terminally differentiated B cells. CD40 is also present on some epithelial cells, carcinomas and lymphoid dendritic cells. This antibody is useful for studies of B-cell growth and function. Anti-CD40 antibodies have been reported to stimulated B-cell proliferation when costimulated with anti-?, anti-CD20 antibodies or with phorbol esters. 5C3 is capable of inducing B-cell proliferation when presented with IL-4. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Especificaciones
CD40 | |
Monoclonal | |
1.0mg/mL | |
TNFRSF5; TNF receptor superfamily member 5; CD40L receptor; Bp50; p50 | |
Mouse | |
0.5 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
5C3 | |
Unconjugated | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.