missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Monoclonal Antibody (145-2C11), Biotin, Invitrogen™
Armenian Hamster Monoclonal Antibody
Marca: Invitrogen MA517656
Detalles adicionales : Peso : 0.01000kg
Descripción
CD3e Monoclonal antibody specifically detects CD3e in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Western Blot.Especificaciones
CD3e | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P04235, P11942, P22646, P24161 | |
Cd247, Cd3d, CD3E, CD3g | |
H-2Kb sp | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
145-2C11 | |
Biotin | |
CD3g | |
4930549J05Rik; A430104F18Rik; AI504783; antigen CD3D, delta polypeptide (TiT3 complex); antigen CD3E, epsilon polypeptide (TiT3 complex); antigen CD3G, gamma polypeptide; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; CD3; CD3 antigen delta chain; CD3 antigen delta polypeptide; CD3 antigen gamma chain; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; CD3 epsilon chain; CD3 epsilon subunit; CD3 epsilon subunit precursor; CD3 gamma-chain; CD3 glycoprotein; CD3 glycoprotein precursor; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3 protein; CD3 TCR complex; CD3 type I transmembrane glycoprotein; CD3 type I transmembrane glycoprotein precursor; CD3 zeta chain; Cd3d; CD3D antigen d | |
Armenian Hamster | |
Protein G | |
RUO | |
12500, 12501, 12502, 12503 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |