missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI2B Monoclonal Antibody (MEM-216), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA110212
Detalles adicionales : Peso : 0.01000kg
Descripción
EVI2B Monoclonal antibody specifically detects EVI2B in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Especificaciones
EVI2B | |
Monoclonal | |
1.0 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P34910 | |
EVI2B | |
Raji cells | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunoprecipitation | |
MEM-216 | |
Unconjugated | |
EVI2B | |
CD361; D17S376; ecotropic viral integration site 2B; ecotropic viral integration site 2B protein; Ecotropic viral integration site 2B protein homolog; EVDB; Evi2; Evi-2; EVI2B; EVI-2B; Protein EVI2B; protein EVI2B-like | |
Mouse | |
Protein A | |
RUO | |
2124 | |
4° C, do not freeze | |
Liquid |