missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30 Monoclonal Antibody (MEM-268), APC, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA119767
Detalles adicionales : Peso : 0.01000kg
Descripción
CD30 Monoclonal antibody specifically detects CD30 in Human samples. It is validated for Flow Cytometry.Especificaciones
CD30 | |
Monoclonal | |
APC | |
TNFRSF8 | |
Ber-H2; Cd30; CD30 antigen; CD30 molecule; CD30L receptor; cytokine receptor CD30; D1S166E; Ki; Ki-1; Ki-1 antigen; Lymphocyte activation antigen CD30; sCD30; soluble CD30; TNF receptor superfamily member 8; TNFRSF8; Tumor necrosis factor receptor superfamily member 8; tumor necrosis factor receptor superfamily, member 8; Tumor necrosis factor superfamily member 8; tumor necrosis factor superfamily, member CD30 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
943 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-268 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P28908 | |
TNFRSF8 | |
Expression vector containing CD30 cDNA (booster suspension of THP-1 cell line). | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG |