missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD275 (B7-H2) Mouse anti-Human, Clone: GM13C1, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA117756
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody recognizes native human ICOSL protein expressed in transfected mammalian cells.
Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.Especificaciones
CD275 (B7-H2) | |
Monoclonal | |
1 mg/mL | |
ICOSLG | |
B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, KIAA0653, LICOS, ICOS Ligand | |
Mouse | |
Protein G | |
RUO | |
23308 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry | |
GM13C1 | |
Unconjugated | |
O75144 | |
ICOSLG | |
cDNA encoding human ICOSL | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |