Learn More
CD26 Monoclonal Antibody (ICO-147), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA17651
Detalles adicionales : Peso : 0.01000kg
Descripción
MA1-7651 detects CD26 from human samples. MA1-7651 has been successfully used in Immunoprecipitation and ELISA procedures. The MA1-7651 immunogen is activated T, B lymphocytes and macrophages.
CD26 Monoclonal antibody specifically detects CD26 in Human samples. It is validated for ELISA, Immunoprecipitation.Especificaciones
CD26 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P27487 | |
DPP4 | |
Activated T, B lymphocytes and macrophages. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation | |
ICO-147 | |
Unconjugated | |
DPP4 | |
ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10 | |
Mouse | |
Protein A | |
RUO | |
1803 | |
4°C short term, -80°C long term | |
Liquid |