Learn More
CD3z Monoclonal Antibody (H146-968), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Marca: Invitrogen MA517673
Detalles adicionales : Peso : 0.01000kg
Descripción
MA5-17673 targets CD247/CD3z in FACS and IF applications and shows reactivity with Mouse samples. The MA5-17673 immunogen is kLH conjugated mouse zeta-chain (CD3) sequence 151-164. MA5-17673 isotype is IgG2, kappa.
CD3z Monoclonal antibody specifically detects CD3z in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.Especificaciones
CD3z | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P24161 | |
CD247 | |
KLH conjugated mouse zeta-chain (CD3) sequence 151-164 | |
300 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2 κ |
Flow Cytometry, Immunocytochemistry | |
H146-968 | |
FITC | |
CD247 | |
4930549J05Rik; A430104F18Rik; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; Cd3; CD3 antigen, zeta polypeptide; CD3 zeta; CD3 zeta chain; CD3e antigen; CD3-epsilon; Cd3-eta; CD3H; CD3Q; Cd3z; CD3Z antigen, zeta polypeptide (TiT3 complex); Cd3zeta; Cd3-zeta; CD3zeta chain; CD3-zeta/eta; FLJ18683; IMD25; T3E; T3z; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor CD3 subunit zeta; T-cell receptor CD3, subunit zeta; T-cell receptor T3 eta chain; T-cell receptor T3 zeta chain; T-cell receptor zeta chain; T-cell surface glycoprotein CD3 zeta chain; TCR zeta chain; TCRE; Tcrk; TCRZ; TCRzeta | |
Armenian Hamster | |
Protein G | |
RUO | |
12503 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |