missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD235a Polyclonal Antibody, FITC, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA535002
Detalles adicionales : Peso : 0.01000kg
Descripción
PA5-35002 targets CD235a/Glycophorin A in FACS applications and shows reactivity with Human samples. The PA5-35002 immunogen is human Glycophorin A transmembrane protein.
CD235a Polyclonal antibody specifically detects CD235a in Human samples. It is validated for Flow Cytometry.Especificaciones
CD235a | |
Polyclonal | |
FITC | |
GYPA | |
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha | |
Rabbit | |
Protein G | |
RUO | |
2993 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Flow Cytometry | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P02724 | |
GYPA | |
Human Glycophorin A transmembrane protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |