Learn More
CD229 Monoclonal Antibody (HLy9.25), APC, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA110150
Detalles adicionales : Peso : 0.01000kg
Descripción
CD229 Monoclonal antibody specifically detects CD229 in Human samples. It is validated for Flow Cytometry.Especificaciones
CD229 | |
Monoclonal | |
APC | |
Ly9 | |
AI893573; CD229; CDABP0070; Cell surface molecule Ly-9; hly9; Lgp100; LY9; Ly-9; Lymphocyte antigen 9; mLY9; RP11-312J18.1; Signaling lymphocytic activation molecule 3; SLAM family member 3; SLAMF3; T100; T-lymphocyte surface antigen Ly-9 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
4063 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HLy9.25 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q9HBG7 | |
Ly9 | |
CD299-transfected 300.19 pre-B cell line | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |