missing translation for 'onlineSavingsMsg'
Learn More

IGF2R Monoclonal Antibody (MEM-238), Alexa Fluor™ 647, Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA518132

245.34 EUR hasta el 2024-03-29
Use el código promocional "21615" para aplicar la oferta.



Detalles adicionales : Peso : 0.01000kg

Código de producto. 15397394

  • 368.00€ / 100 tests
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody recognizes an epitope between domains 2 and 5 of CD222 (IGF2 receptor), a ubiquitously expressed 250 kDa multifunctional type I transmembrane protein. The majority of CD222 is found in the late endosomal/prelysosomal compartment, 5-10% in the plasma membrane and the truncated (220 kDa) form of CD222 is present in human and bovine serum.

IGF2R Monoclonal antibody specifically detects IGF2R in Human, Non-human Primate samples. It is validated for Flow Cytometry, Western Blot.
Especificaciones

Especificaciones

IGF2R
Monoclonal
Alexa Fluor 647
IGF2R
300 kDa mannose 6-phosphate receptor; AI661837; cation-independent mannose 6-phosphate receptor; cation-independent mannose 6-phosphate receptor protein; cation-independent mannose-6-phosphate receptor; cation-independent MPR; CD222; CI Man-6-P receptor; CI-M6PR; CIMPR; CI-MPR; Igf2r; IGF-II receptor; IGF-II/CI-MPR; Igfr2; insulin like growth factor 2 receptor; insulin-like growth factor 2 receptor; Insulin-like growth factor II receptor; insulin-like growth factor receptor type 2; M6P; M6P/IGF2 receptor; M6P/IGF2R; M6PR; M6P-R; mannose-6-phosphate receptor, cation independent; mannose-6-phosphate/insulin-like growth factor II receptor; MPR 300; MPR1; MPR300; MPRI; similar to mannose-6-phosphate/insulin-like growth factor II receptor
Mouse
Size-exclusion chromatography
RUO
3482
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry, Western Blot
MEM-238
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P11717
IGF2R
Recombinant Vaccinia virus encoding CD222
100 Tests
Primary
Human, Non-human Primate
Antibody
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales