missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD206 Monoclonal Antibody (7-450), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA516546
Detalles adicionales : Peso : 0.01000kg
Descripción
A suggested positive control for immunohistochemical applications is human tonsil.
CD206 Monoclonal antibody specifically detects CD206 in Human samples. It is validated for Immunohistochemistry (Frozen).Especificaciones
CD206 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P22897 | |
Mrc1 | |
Monocyte-derived dendritic cells. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Frozen) | |
7-450 | |
Unconjugated | |
Mrc1 | |
AW259686; bA541I19.1; CD206; CELE_Y39A3CR.4; CLEC13D; CLEC13DL; C-type lectin domain family 13 member D; C-type lectin domain family 13 member D-like; ddp-1; hMR; Human mannose receptor; macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1; mannose receptor, C type 1; mannose receptor, C type 1-like 1; Mitochondrial import inner membrane translocase subunit Tim8; MMR; MR; MRC1; MRC1L1; tim-8; Y39A3CR.4 | |
Mouse | |
Protein A | |
RUO | |
4360 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |