Learn More
CD181 Mouse anti-Human, APC, Clone: 5A12, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 551080
Detalles adicionales : Peso : 0.02000kg
Descripción
Reacts with Interleukin-8 receptor A (IL-8RA), a G protein-linked seven transmembrane domain protein, expressed on monocytes and granulocytes. It is also expressed on a subset of T cells. Both IL-8RA and IL-8RB are high affinity receptors for IL-8. IL-8 is an inflammatory cytokine which induces chemotaxis of neutrophils, basophils and T lymphocytes. IL-8RA and IL-8RB were first clustered as CD128a and CD128b, respectively in the VIIth HLDA workshop,and then were re-clustered as CD181 and CD182, respectively in the VIIIth HLDA workshop.
Especificaciones
CD181 | |
Monoclonal | |
APC | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
5A12 | |
CXCR1, IL-8RA, CD128a | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG2b κ |
For Research Use Only.