Learn More
CD100 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA14202
Detalles adicionales : Peso : 0.01000kg
Descripción
PA1-4202 detects CD100 in human samples. PA1-4202 has been successfully used in Western blot procedures. By Western blot this antibody detects a ∽158 kDa protein representing CD100 in transfected CHO cells. The PA1- 4202 immunogen is a synthetic peptide corresponding to residues K(777) P K S D F C D R E Q S(788) of human CD100.
CD100 Polyclonal antibody specifically detects CD100 in Human samples. It is validated for Western Blot.Especificaciones
CD100 | |
Polyclonal | |
Unconjugated | |
Sema4d | |
A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10507 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q92854 | |
Sema4d | |
Synthetic peptide corresponding to residues K(777) P K S D F C D R E Q S(788) of human CD100. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |