missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 3 Monoclonal Antibody (74T2), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen 437800
Detalles adicionales : Peso : 0.50000kg
Descripción
Caspase 3 Monoclonal antibody specifically detects Caspase 3 in Equine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Especificaciones
Caspase 3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P42574, P55213, P70677 | |
CASP3 | |
Recombinant protein derived from the N-terminus of human Caspase 3 protein | |
100 μg | |
Primary | |
Equine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
74T2 | |
Unconjugated | |
CASP3 | |
A830040C14Rik; AC-3; Apopain; Apopain precursor; Casp3; CASP-3; caspase 3; Caspase 3 apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis-related cysteine peptidase; caspase 3, apoptosis-related cysteine protease; Caspase3; caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; Caspase-3-like protein; CC3; Cpp32; CPP-32; CPP32B; CPP32beta; cysteine protease CPP32; cysteine protease; CPP32; apopain; ICE3; IRP; LICE; mLdy; PARP cleavage protease; procaspase3; protein Yama; SCA-1; SREBP cleavage activity 1; Yama; yama protein | |
Mouse | |
Protein A | |
RUO | |
100034083, 12367, 25402, 836 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |