missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 2 Monoclonal Antibody (18E809.3), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA141315
Detalles adicionales : Peso : 0.01000kg
Descripción
Suggested positive control: HL-60.
Caspase 2 Monoclonal antibody specifically detects Caspase 2 in Human samples. It is validated for Western Blot.Especificaciones
Caspase 2 | |
Monoclonal | |
Conc. Not Determined | |
ascites diluted in PBS with 0.2% gelatin and 0.05% sodium azide | |
P42575 | |
CASP2 | |
A peptide corresponding to amino acids 400-450 of human Caspase-2 | |
RUO | |
835 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
18E809.3 | |
Unconjugated | |
CASP2 | |
CASP2; CASP-2; caspase 2; caspase 2 apoptosis-related cysteine peptidase; caspase-2; Caspase-2 subunit p12; Caspase-2 subunit p13; Caspase-2 subunit p18; ICH1; ICH-1; ICH-1 protease; ICH-1L; ICH-1L/1S; NEDD2; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; PPP1R57; Protease ICH-1; protein phosphatase 1, regulatory subunit 57 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |