missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA520297
Detalles adicionales : Peso : 0.01000kg
Descripción
A suggested positive control is Jurkat cell lysate. PA5-20297 can be used with blocking peptide PEP-0407.
Caspase 14 Polyclonal antibody specifically detects Caspase 14 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
Caspase 14 | |
Polyclonal | |
Unconjugated | |
Casp14 | |
apoptosis-related cysteine protease; CASP14; CASP-14; caspase 14; caspase 14, apoptosis-related cysteine peptidase; caspase 14, apoptosis-related cysteine protease; Caspase14; caspase-14; Caspase-14 subunit p10, mature form; Caspase-14 subunit p17, mature form; Caspase-14 subunit p20, intermediate form; Caspase-14 subunit p8, intermediate form; caspase-14; LOW QUALITY PROTEIN: caspase-14; MICE; Mini-ICE | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12365, 23581, 299587 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O89094, P31944 | |
Casp14 | |
A 16 amino acid peptide from near the carboxy terminus of human caspase-14. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |