Learn More
CASK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca:  Invitrogen PA517433
Detalles adicionales : Peso : 0.01000kg
Código de producto. 11566251
  • 422.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock
Descripción
Descripción
It is not recommended to aliquot this antibody.
CASK Polyclonal antibody specifically detects CASK in Human, Mouse, Rat samples. It is validated for Western Blot.
Especificaciones
Especificaciones
CASK
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
O14936, O70589, Q62915
Cask
Synthetic peptide corresponding to the amino-terminus of CASK
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
Unconjugated
Cask
CAGH39; calcium/calmodulin dependent serine protein kinase; calcium/calmodulin-dependent serin protein kinase; Calcium/calmodulin-dependent serine protein kinase; calcium/calmodulin-dependent serine protein kinase (MAGUK family); calcium/calmodulin-dependent serine protein kinase membrane-associated guanylate kinase; CAMGUK; CASK; CMG; DXPri1; DXRib1; FGS4; G-protein-coupled receptor GPR34; hCASK; LIN2; LIN-2; MGC7449; MICPCH; mLin-2; MRXSNA; OTTHUMP00000023157; OTTHUMP00000023161; OTTHUMP00000023162; Pals3; peripheral plasma membrane protein CASK; Protein lin-2 homolog; RP23-278L4.1; TNRC8; trinucleotide repeat containing 8
Rabbit
Antigen affinity chromatography
RUO
12361, 29647, 8573
-20°C