Learn More
Annexin I Mouse, Unlabeled, Clone: 29, BD
Mouse Monoclonal Antibody
382.00€ - 565.00€
Especificaciones
Antígeno | Annexin I |
---|---|
Clon | 29 |
Concentración | 250μg/mL |
Aplicaciones | Western Blot |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15815239
|
BD Biosciences
610066 |
50 μg |
382.00€
1 set |
Fecha estimada de envío: 21-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15825239
|
BD Biosciences
610067 |
150 μg |
565.00€
1 set |
Fecha estimada de envío: 21-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Annexin I, also known as lipocortin-1 or calpactin II, is part of a calcium-dependent phospholipid-binding protein family. Members share a common core domain, but each has a unique N-terminal tail that imparts its functional specificity. It is believed that phosphorlyation of this region in annexin I regulates its functions. Annexin I has been implicated in regulating phospholipid vesicle aggregation, mediating inflammatory response, and inhibiting the activity of phospholipase A2. Additionally, it has been found that the 10kDa protein S100C influences annexin I's activity by binding to the N-terminal 13 residues of annexin in a manner similar to that of the annexin II/p11 complex.
Host Species: Mouse
Clone: 29
Isotype: IgG1
Species Reactivity: Human
Immunogen: Cow Annexin I aa. 1-346
Formula Weight [Chemical]: 38kDa
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Especificaciones
Annexin I | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Lipocortin-1; Calpactin II |
29 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human | |
Cow Annexin I aa. 1-346 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.