Learn More
Adaptin δ Mouse anti-Human, Unlabeled, Clone: 18, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 611329
Detalles adicionales : Peso : 0.01000kg
Descripción
Sorting of integral membrane proteins at various stages of the endocytic and secretory pathways is mediated by vesicular trafficking between a variety of organelles. Two sorting signals are tyrosine-based and dileucine-based signals that interact with heterotetrameric adaptor protein complexes (AP-1 through AP-4), which are associated with the vesicle coats. These coatomers contain two large adaptin proteins (γ, α, δ, ε and β1, β2, β3, β4, respectively) that are noncovalently linked to one medium chain (μ1, μ2, μ3, or μ4) and one small chain (σ1, σ2, σ3, or σ4). The AP-1 and AP-3 complexes are involved in protein sorting from the TGN and endosomes, while AP-2 adaptor complexes are involved in clathrin-mediated endocytosis. In the AP-3 complex, adaptin δ contains an N-terminal “head” domain that has 25% identity with adaptins α and γ, while its C-terminal “hinge” and “ear” domains are two-fold larger and more divergent as compared to the same domains in adaptins α and γ. Adaptin δ is widely expressed and its mutation results in protein storage deficiencies within lysosomal-related organelles in platelets, melansomes, and synaptic vesicles
Host Species: Mouse
Clone: 18
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Adaptin δ aa. 627-731
Formula Weight [Chemical]: 160kDa
Immunofluorescence, Western Blotting
Especificaciones
Adaptin δ | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human Adaptin δ aa. 627-731 | |
150 μg | |
Cell Biology | |
Human | |
IgG1 |
Western Blot | |
18 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.