missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine Receptor α Mouse, Unlabeled, Clone: 26, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 610988

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15825749

  • 251.00€ / 1 set


Fecha estimada de envío
09-12-2019
Añadir a la cesta

Descripción

Descripción

Acetylcholine is an amine neurotransmitter at the neuromuscular junction. It is released from the presynaptic membrane of a cholinergic synapse into the synaptic cleft. It diffuses across the cleft and binds acetylcholine receptors (AChR) on the postsynaptic membrane. Receptor binding induces postsynaptic membrane depolarization and the generation of an action potential that produces effects such as muscle contraction. The AChR is a 250kDa pentameric complex of four transmembrane subunits in a stoichiometry of α2βγδ. In response to ligand binding, all subunits participate in the formation of an integral cation channel. However, the acetylcholine binding site is primarily within the α subunit. Myasthenia gravis (MG) is an autoimmune condition in which AchR levels are decreased. Autoantibodies bind and crosslink the AchRs leading to their internalization and degradation. This results in a decreased number of functional AChRs. Patients develop muscular weakness and some voluntary muscle fatigue. However, development of MG is also affected by genetic factors. One of the allelic forms of the AChRα gene appears to significantly contribute to MG susceptibility.

Immunofluorescence, Western Blotting

Especificaciones

Especificaciones

Acetylcholine Receptor α
26
Unconjugated
Affinity Purified
Rat Acetylcholine Receptor α aa. 332-457
50μg
Cell Biology
Primary
Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Videos
Documentos

For Research Use Only.