Learn More
Ansell Vantage™ Cut-Resistant Gloves
Click to view available options
Medium-weight; 10 gauge; With Dyneema-based yarn; Size 9
Descripción
Revolutionary engineered yarns provide increased cut protection without the bulk. Ideal for work environments where sharp parts and materials are handled.
- Blended fiber, lightweight construction provides all-day comfort
- Contour design offers flexibility for reduced hand fatigue
- Medium weight, 10-gauge yarn with 100% Kevlar™ aramid fiber reinforced thumb crotch
Especificaciones
Cut Resistance Rating | ANSI A4 |
Material | Dyneema™ |
Tipo de producto | Cut-resistant Gloves |
Tamaño | 9 |
Para utilizar con (aplicación) | Appliance manufacturing, automotive, general carpentry, general industry, glass, masonry/tile, metal fabrication, metalworking, mining, plumbers, textiles |
Estilo de puño de los guantes | Knit |