Learn More
Ansell Vantage™ Cut-Resistant Gloves
Catalog Number 12666961
Click to view available options
Medium-weight; 10 gauge; With Dyneema-based yarn; Size 7
Revolutionary engineered yarns provide increased cut protection without the bulk. Ideal for work environments where sharp parts and materials are handled.
  • Blended fiber, lightweight construction provides all-day comfort
  • Contour design offers flexibility for reduced hand fatigue
  • Medium weight, 10-gauge yarn with 100% Kevlar™ aramid fiber reinforced thumb crotch
Cut Resistance Rating ANSI A4
Material Dyneema™
Tipo de producto Cut-resistant Gloves
Tamaño 7
Para utilizar con (aplicación) Appliance manufacturing, automotive, general carpentry, general industry, glass, masonry/tile, metal fabrication, metalworking, mining, plumbers, textiles
Estilo de puño de los guantes Knit