missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 58-530 Nitrile Chemical Resistant Gloves

Ideal for incidental exposure to lubricants and cleaning agents in industrial settings. Ansell™ AlphaTEC™ Gloves protects against active chemical contact in refining, petrochemical and related industries.

64.45€

Especificaciones

Color Black, Red
Material Nitrile
Ver más especificaciones

Productos 1
Código de producto Marca TamañoTamaño Precio Cantidad y disponibilidad  
Código de producto Marca TamañoTamaño Precio Cantidad y disponibilidad  
12465000 Ver documentos Ansell™
58530B090
9 64.45€
6 pares Ver productos alternativos
Ahorre hasta 
»

Stock Remoto 14
Añadir a la cesta
 
Descripción

Descripción

  • Liquid and chemical protection gloves
  • Incorporates revolutionary Ansell Grip Technology™ to enable users to handle wet or oily objects with less grip force and more control
  • Unique combination of liquid-proof chemical resistance and grip, together with flexibility and dexterity, makes the logical choice when working with chemicals
  • Reliable chemical protection for added user safety
  • Patented Ansell Grip Technology™ ensures that the coating polymer does not penetrate the liner during manufacture
  • Coating is therefore consistent and sound, significantly reducing the likelihood of chemical leakage onto the skin
  • AlphaTec™ models with white nylon liner are recommended for indoor applications, whereas with black acrylic liner are recommended for outdoor applications

Applications:

  • Applying sealers, touching up
  • Body painting including surface inspection
  • Changing tools and dies
  • Handling application and cleaning tools
  • Loading and unloading process equipment
  • Metalworking and assembly operations
  • Opening furnaces draining pumps valves or lines and crackers BTX process
  • Painting
  • Painting or cleaning tools or robots
  • Transferring liquids and solids between vessels and tanks and process equipment
  • Warehousing of raw materials
Especificaciones

Especificaciones

Black, Red
Nitrile