missing translation for 'onlineSavingsMsg'
Learn More

Annexin V Mouse anti-Human, Mouse, Clone: 1E6A8, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66245-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817945

  • 319.00€ / 150 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
Especificaciones

Especificaciones

Annexin V
1E6A8
Unconjugated
ANXA5
Liquid
Anxa5
Annexin V Fusion Protein Ag1538
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.93 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08758, P48036
Anchorin CII, Annexin 5, annexin A5, Annexin V, ANX5, ANXA5, Calphobindin I, CBP I, Endonexin II, ENX2, Lipocortin V, PAP I, PP4, Thromboplastin inhibitor, VAC alpha, Vascular anticoagulant alpha
Mouse
IgG1
150 μL
-20°C
Primary
11747, 308
Videos
SDS
Documentos
Certificados