missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11427-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This is a rabbit polyclonal antibody produced by the full-length ANKRD1 of human origin.
The protein encoded by this gene is localized to the nucleus of endothelial cells and is induced by IL-1 and TNF-alpha stimulation. Studies in rat cardiomyocytes suggest that this gene functions as a transcription factor. Interactions between this protein and the sarcomeric proteins myopalladin and titin suggest that it may also be involved in the myofibrillar stretch-sensor system.Especificaciones
ANKRD1 | |
Polyclonal | |
Unconjugated | |
ANKRD1 | |
ALRP, ANKRD1, C 193, C193, Cardiac ankyrin repeat protein, CARP, CVARP, HA1A2, MCARP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107765, 27063, 27064 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15327, Q8R560, Q9CR42 | |
Ankrd1 | |
ANKRD1 Fusion Protein Ag1970 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |