missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiotensinogen Mouse anti-Human, Clone: 2H2A10, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60126-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
AGT, pre-angiotensinogen or angiotensinogen precursor, is expressed in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. The resulting product, angiotensin I, is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. The protein is involved in maintaining blood pressure and in the pathogenesis of essential hypertension and preeclampsia. Mutations in AGT gene are associated with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. Defects in AGT gene have also been associated with non-familial structural atrial fibrillation, and inflammatory bowel disease.Especificaciones
Angiotensinogen | |
Monoclonal | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01019 | |
AGT | |
Angiotensinogen Fusion Protein Ag2632 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
2H2A10 | |
Unconjugated | |
AGT | |
AGT, Ang I Angiotensin 2, Ang II Angiotensin 3, Ang III, Angiotensin, Angiotensin I, Angiotensin II, Angiotensin III, Angiotensinogen, ANHU, Des Asp[1] angiotensin II, Serpin A8, SERPINA8 | |
Mouse | |
Protein A | |
RUO | |
183 | |
-20°C | |
Liquid |