missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin-2 Antibody (117), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89600V

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213939

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Description

Description

Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Angiopoietin-2
Monoclonal
DyLight 405
AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand
This antibody was obtained from a rabbit immunized with purified, recombinant Human Angiopoietin-2 (NP_001138.1; Met1-Phe496).
0.1 mL
Angiogenesis, Cancer
285
Store at 4°C in the dark.
IgG
Western Blot, Immunofluorescence
117
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers