missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ANG (Human) Recombinant Protein
Human ANG full-length ORF ( AAH54880, 25 a.a. - 147 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000283-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: QDNSRYTHFLTQHYDAKPQGRDDRYCESIMRRRGLTSPCKDINTFIHGNKRSIKAICENKNGNPHRENLRISKSSFQVTTCKLHGGSPWPPCQYRATAGFRNVVVACENGLPVHLDQSIFRRP
Especificaciones
AAH54880 | |
angiogenin, ribonuclease, RNase A family, 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANG | |
GST |
283 | |
Wheat germ expression system | |
25 μg | |
ALS9, HEL168, MGC22466, MGC71966, RNASE4, RNASE5 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |