Learn More
Abeba™ Anatom 32268 Shoes
Product Code. 15649567
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15649567 43 1 pair
15679557 36 1 pair
15689557 37 1 pair
15699557 38 1 pair
15609567 39 1 pair
15619567 40 1 pair
15629567 41 1 pair
15639567 42 1 pair
15659567 44 1 pair
15669567 45 1 pair
15679567 46 1 pair
15689567 47 1 pair
15699567 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 15649567 Supplier Abeba™ Supplier No. 32268/43
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S3 SRC
  • Smooth leather
  • Black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Metalfree penetration resistance
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Scratch-resistant coating in toe and heel area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S3 SRC
Color Black
Material Leather
Size 43
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32268
Toe Material Steel