missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluoroborato de amonio, 99,999 % (base metálica), Thermo Scientific Chemicals

Catalog Number p-7059127
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
13826-83-0
BF4H4N
104.84
MFCD06245785
PDTKOBRZPAIMRD-UHFFFAOYSA-O
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
9964072
azanio;tetrafluoroborato
[NH4+].F[B-](F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
13826-83-0
BF4H4N
104.84
MFCD06245785
PDTKOBRZPAIMRD-UHFFFAOYSA-O
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
9964072
azanio;tetrafluoroborato
[NH4+].F[B-](F)(F)F

Ammonium fluoroborate is used as an analytical reagent. It is also applied in the textile printing and dyeing industry and used as a catalyst for resin finishing. It is mainly used for the high temperature fluxing action required by the metals industry; catalyst; in flame retardants and acts as a solid lubricant in cutting-oil emulsions for aluminum rolling and forming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ammonium tetrafluoroborate
Punto de fusión 220°C (sublimation)
Color Blanco
Forma física cristalino/a/s
Cantidad 5 g
Notas del porcentaje del ensayo (base metálica)
Número UN UN3260
Información de solubilidad Soluble in water at 16°C 25g/L.
Formula Weight (peso de la fórmula) 104.84
Porcentaje de pureza 99.999%
Olor Ammonia-like
Sensibilidad Hygroscopic
Densidad 1.871 g/mL
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'