Learn More
Ammonium iron(III) sulfate dodecahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-102.0%, crystalline, slightly violet, Honeywell Fluka™
Marca: Honeywell Fluka 31111-2.5KG
Detalles adicionales : CAS : 7783-83-7 Peso : 2.50000kg
Cantidad. | 2.5kg |
---|
Identificadores de productos químicos
7783-83-7 | |
99 to 102% | |
482.19g/mol | |
NH4Fe(SO4)2 · 12H2O | |
Reag. Ph. Eur. reag. ISO | |
In accordance with water, 10g/dL |
Ammonium Iron(Iii) Sulfate Dodecahydrate | |
Slightly Violet | |
puriss. p.a. ACS Reagent | |
MFCD00150004 | |
Crystalline | |
NONH for all modes of transport |
Especificaciones
Ammonium Iron(Iii) Sulfate Dodecahydrate | |
7783-83-7 | |
99 to 102% | |
482.19g/mol | |
NONH for all modes of transport | |
Crystalline | |
2.5kg |
Reag. Ph. Eur. reag. ISO | |
puriss. p.a. ACS Reagent | |
NH4Fe(SO4)2 · 12H2O | |
MFCD00150004 | |
In accordance with water, 10g/dL | |
Slightly Violet |