missing translation for 'onlineSavingsMsg'
Learn More

Aminopeptidase LRAP/ERAP2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-81783G

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213288

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Aminopeptidase LRAP/ERAP2 Polyclonal antibody specifically detects Aminopeptidase LRAP/ERAP2 in Human samples. It is validated for Western Blot, ELISA
Especificaciones

Especificaciones

Aminopeptidase LRAP/ERAP2
Polyclonal
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA
DyLight 488
EC 3.4.11, EC 3.4.11.3, endoplasmic reticulum aminopeptidase 2, FLJ23633, FLJ23701, FLJ23807, Leukocyte-derived arginine aminopeptidase, LRAPL-RAPEC 3.4.11.-
ERAP2 antibody was raised against a 17 amino acid peptide near the amino terminus of human ERAP2. The immunogen is located within amino acids 110 - 160 of ERAP2 . Amino Acid Squence: LQSEEDSRYMKPGKELK
0.1 mL
Innate Immunity
64167
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales